Learn More
Strem, An Ascensus Company CAS# 7646-85-7. 10g. Zinc chloride (99.99%-Zn) PURATREM. MFCD00011295
SDP
Supplier:  Strem, An Ascensus Company 93305210G
Encompass
CAS# 7646-85-7. 10g. Zinc chloride (99.99%-Zn) PURATREM. MFCD00011295. Molecular Weight: 136.28. Molecular Formula: ZnCl2. Color/form: -10 mesh spheres Strem# 93-3052. http://www.strem.com/catalog/v/93-3052/
Catalog No. 50-901-15005