Learn More
Medchemexpress LLC Zinc dihydrogen phosphate | 13598-37-3 | MFCD00152955 | 98.0% | 261.39 g/mol | H3O4P·1/2Zn | 100 G

Supplier: Medchemexpress LLC HYW414898100G
Zinc dihydrogen phosphate is an inorganic phosphate salt provided as a high-purity reagent for laboratory and research use. It is typically used as an analytical reagent and preservative in life-science applications and is characterized by CAS 13598-37-3 and a molecular weight of 261.39 g/mol.
- High purity (98.0%) suitable for analytical applications.
- Useful as an analytical reagent and preservative.
- Available in common lab pack sizes including 100 g.
- Molecular weight 261.39 g/mol and CAS number 13598-37-3 for reference.
- Store in a cool, dry place and protect from moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.