Learn More
Medchemexpress LLC Zinc dihydrogen phosphate | 13598-37-3 | MFCD00152955 | 98.0% | 261.39 g/mol | Zn(H2PO4)2 | 1 KG

Supplier: Medchemexpress LLC HYW4148981KG
Zinc dihydrogen phosphate is an inorganic zinc phosphate supplied as a high-purity solid for use as an analytical reagent and preservative in life-science research, formulation studies, and materials chemistry. It is commonly used where a soluble zinc phosphate source is required for analytical or formulation applications.
- High purity (98.0%) suitable for analytical work.
- CAS number 13598-37-3 for unambiguous identification.
- Chemical formula Zn(H2PO4)2 for phosphate chemistry applications.
- Available as a 1 kg pack for laboratory and bulk use.
- Water and acid soluble zinc source compatible with phosphate-containing systems.
- Stable solid form for convenient handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.