Learn More
Medchemexpress LLC Zinc dihydrogen phosphate | 13598-37-3 | MFCD00152955 | 98.0% | 261.39 | H3O4P.1/2Zn | 500 G
SDP
Supplier:  Medchemexpress LLC HYW414898500G
Encompass_Preferred
Zinc dihydrogen phosphate is an inorganic zinc phosphate reagent used as an analytical reagent and preservative in laboratory and life-science research. It typically appears as a white crystalline solid, is water soluble, and is supplied with a nominal purity of 98% and a molecular weight of 261.39 g/mol. It is suitable for biochemical assays, formulation studies, and phosphate chemistry applications.
  • Used as an analytical reagent and preservative in laboratory workflows.
  • High nominal purity (98%) for consistent results.
  • Water-soluble white crystalline solid suitable for solution preparation.
  • Applicable in biochemical assays and formulation development.
  • Available in multiple pack sizes for flexible laboratory use.
Catalog No. 50-002-81064