Learn More
Medchemexpress LLC Zinc oxide 100 nm 10g
SDP
Supplier:  Medchemexpress LLC HYW116336D10G
Encompass_Preferred
Zinc Oxide 100 nm particle size can be used to prepare biomaterials and for sensor research
Catalog No. 50-002-06670