Learn More
Strem, An Ascensus Company CAS# 1314-13-2. 500g. Zinc oxide, 99.7%. MFCD00011300
SDP
Supplier:  Strem, An Ascensus Company 933017500G
Encompass
CAS# 1314-13-2. 500g. Zinc oxide, 99.7%. MFCD00011300. Molecular Weight: 81.37. Molecular Formula: ZnO. Color/form: white pwdr. (mean particle size 0.31 microns) Strem# 93-3017. http://www.strem.com/catalog/v/93-3017/
Catalog No. 50-901-14972