Learn More
Medchemexpress LLC Zinc Phytate | 63903-51-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN25805MG
Encompass_Preferred
Zinc Phytate (myo-Inositol; hexakis dihydrogen phosphate; Inositol hexaphosphate) is an orally active compound derived from legume seeds. It functions as a [PO4]3- storage depot and a precursor for other inositol phosphates and pyrophosphates. It is used in research related to cardiovascular disease, metabolic disease, nervous system disease, and cancer.
  • Attenuates Aβ oligomers
  • Upregulates autophagy protein
  • Orally active compound
  • Derived from legume seeds
  • Precursor for inositol phosphates and pyrophosphates
Catalog No. 50-004-20294