Learn More
Cayman Chemical ZInc ProtoporphyrIn-9 10mg
SDP
Supplier:  Cayman Chemical 1448310 MG
Encompass_Preferred
A competitive inhibitor of heme oxygenase that also inhibits soluble guanylyl cyclase and inactivates the NOS isoforms nNOS, iNOS, and eNOS (IC50s = 0.8, 4, and 5 μM, respectively); used to assess iron status and to diagnose iron disorders and has shown therapeutic potential in hyperbilirubinemia
Catalog No. 50-296-9722