Learn More
Medchemexpress LLC Zinc protoporphyrin (Zn(II)-protoporphyrin IX) | 15442-64-5 | 98.9% | 626.02 g·mol⁻¹ | C34H32N4O4Zn | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10119325MG
Encompass_Preferred
Zinc protoporphyrin (Zn(II)-protoporphyrin IX) is a metalloporphyrin and a competitive heme oxygenase-1 (HO-1) inhibitor used as a research reagent and as a biomarker for iron deficiency. It is employed in studies of oxidative stress, heme metabolism, and cancer biology. Characterization: molecular formula C34H32N4O4Zn; molecular weight 626.02 g·mol⁻1.
  • Competitive heme oxygenase-1 (HO-1) inhibitor.
  • Used as a biomarker for iron deficiency in biological samples.
  • Applicable to studies of oxidative stress, heme metabolism, and cancer biology.
  • Characterized molecular formula C34H32N4O4Zn and molecular weight 626.02 g·mol⁻1.
  • High purity validated by HPLC (~98.9%).
  • Available in small pack sizes suitable for laboratory assays.
Catalog No. 50-002-12950