Learn More
Apexbio Technology LLC Zinc Pyrithione, 1g Cas# 13463-41-7
SDP
Supplier:  Apexbio Technology LLC B22011000
Encompass_Preferred
Zinc pyrithione is an antifungal and antibacterial agent disrupting membrane transport by blocking the proton pump. 13463-41-7. MFCD00067336 Zinc pyrithione is an antifungal and antibacterial agent disrupting MEM transport by blocking the proton pump. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8689