Learn More
Alkali Scientific Zinc Sulfate Heptahydrate, 1 Kg
SDP
Supplier:  Alkali Scientific ZIN001CP
Encompass_Preferred
Zinc Sulfate Heptahydrate, available in a 1kg quantity, is a laboratory-grade chemical compound used in a variety of applications. It is primarily employed in the preparation of buffer solutions and as a source of zinc ions in biochemical experiments. Zinc sulfate plays a critical role in enzyme activation, as many enzymes require zinc as a cofactor. It is also used in agricultural research, where it is applied as a micronutrient for plant growth studies. Zinc sulfate is utilized in clinical and diagnostic labs for research into zinc deficiency and in industrial processes requiring zinc as part of chemical formulations.
Catalog No. 50-006-61755