Learn More
Research Products International Corp Zinc Sulfate Heptahydrate 5KG

Supplier: Research Products International Corp Z221505000.0
CAS Number: 7446-20-0
Molecular Formula: ZnSO4 • 7H2O
Molecular Weight: 287.5
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.