Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc sulfate heptahydrate | 7446-20-0 | 100 G
SDP

Supplier:  Medchemexpress LLC HYN3025100G

Encompass_Preferred

Zinc sulfate heptahydrate is a hydrate, specifically the heptahydrate form of zinc sulfate. It functions as a dietary supplement addressing zinc deficiency and preventing this condition in at-risk individuals.

  • Used as a dietary supplement
  • Addresses zinc deficiency
  • Prevents zinc deficiency in high-risk individuals
  • Purity of 99.8%
  • Appears as a solid
  • Color is white to off-white

Catalog No. 50-004-21250


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.