Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc sulfate heptahydrate | 7446-20-0 | 50 G
SDP

Supplier:  Medchemexpress LLC HYN302550G

Encompass_Preferred

Zinc sulfate heptahydrate is a hydrate that is the heptahydrate form of zinc sulfate. It is a dietary supplement used for zinc deficiency and to prevent the condition in those at high risk. This product is for research use only and not intended for sale to patients.

  • Purity of 99.8%
  • Molecular weight of 287.55
  • Molecular formula: H14O11SZn
  • Solid appearance
  • White to off-white color
  • Soluble in H2O at 100 mg/mL (requires ultrasonic)
  • Store at room temperature, keep dry and cool

Catalog No. 50-004-21316


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.