Learn More
Selleck Chemical LLC Zinc sulfate heptahydrate E2968-50MG
SDP
Supplier:  Selleck Chemical LLC E296850MG
Encompass_Preferred
Zinc sulfate heptahydrate is a hydrate that is the heptahydrate form of zinc sulfate Zinc sulfate heptahydrate is a dietary supplement used for zinc deficiency and can also accelerate the rate of healing It is also used in prepration of zinc tris(thiourea) sulphate (ZTS) a semiorganic nonlinear optic (NLO) material that may be useful in laser fusion experiments and in second-harmonic-generation (SHG) devices
Catalog No. 50-000-40488