Learn More
Alkali Scientific Zinc Sulfate Monohydrate, USP 500 Grams
SDP
Supplier:  Alkali Scientific INC003CP
Encompass_Preferred
Zinc Sulfate Monohydrate, USP Grade (500 g), is a laboratory-grade reagent that is frequently used in pharmaceutical and industrial applications. This compound provides a source of zinc ions, which are essential for various chemical reactions, as well as in the synthesis of other zinc-based compounds. It is commonly used in agriculture as a micronutrient in fertilizers, and in animal feed supplements to address zinc deficiencies. In laboratory settings, it is used for preparation of solutions for analytical tests, medical research, and other scientific studies requiring zinc in precise concentrations.
Catalog No. 50-006-59686