Learn More
Medchemexpress LLC Zinc sulfide | 1314-98-3 | ≥99.0% | 100 G

Supplier: Medchemexpress LLC HYB1821100G
Zinc monosulfide is identified as a biological molecule.
- Appearance: Solid
- Color: White to off-white
- Molecular Weight: 97.45
- Formula: ZnS
- SMILES: [Zn]=S
- Solubility: In Vivo Dissolution Calculator available
- Usage: For research use only. Not sold to patients.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.