Learn More
Medchemexpress LLC Zinc sulfide | 1314-98-3 | ≥99.0% | 25 G

Supplier: Medchemexpress LLC 25GHYB1821
Zinc monosulfide (ZnS) is a biological molecule sold for research purposes only, not for patient use. It has a molecular weight of 97.45 and appears as a white to off-white solid.
- Molecular formula: ZnS
- CAS number: 1314-98-3
- Purity: ≥99.0%
- Appearance: White to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.