Learn More
Strem, An Ascensus Company CAS# 1314-98-3. 100g. Zinc sulfide (99.9%-Zn). MFCD00011301
SDP
Supplier:  Strem, An Ascensus Company 933025100G
Encompass
CAS# 1314-98-3. 100g. Zinc sulfide (99.9%-Zn). MFCD00011301. Molecular Weight: 97.43. Molecular Formula: ZnS. Color/form: white pwdr. Strem# 93-3025. http://www.strem.com/catalog/v/93-3025/
Catalog No. 50-901-14979