Learn More
Strem, An Ascensus Company CAS# 54010-75-2. 5g. Zinc trifluoromethanesulfonate, min. 98% (Zinc triflate). MFCD00013229
SDP
Supplier:  Strem, An Ascensus Company 3040005G
Encompass
CAS# 54010-75-2. 5g. Zinc trifluoromethanesulfonate, min. 98% (Zinc triflate). MFCD00013229. Molecular Weight: 363.54. Molecular Formula: Zn(SO3CF3)2. Color/form: white pwdr. Strem# 30-4000. http://www.strem.com/catalog/v/30-4000/
Catalog No. 50-901-11396