Learn More
Abcam Zinpyr-1, Fluorogenic Zn2+ reporter, 125MG
Supplier:  Abcam AB145349125MG
Encompass_Preferred
MW 823.72 g/mol, Purity >95%. Cell-permeable fluorogenic Zn²+ reporter. 2,7-Dichlorofluorescein (ab146332) derivative. Induces positive fluorescence upon metal ion complexation. PET sensor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3256