Learn More
Medchemexpress LLC Zinquin ethyl ester | 181530-09-6 | >98.0% | 414.5 g·mol⁻¹ | C21H22N2O5S | 1MG
SDP
Supplier:  Medchemexpress LLC HY1241711MG
Encompass_Preferred
Zinquin ethyl ester is a quinolone-based, cell-permeable fluorescent probe for selective detection of intracellular Zn(II). The ethyl ester facilitates membrane penetration and is cleaved by intracellular esterases to trap the active fluorophore, enabling live-cell measurement of cytosolic zinc fluxes and localization. Supplied as a high-purity solid for research use.
  • Cell-permeable probe for intracellular zinc detection.
  • Ethyl ester is cleaved intracellularly to retain signal.
  • Suitable for live-cell fluorescence imaging and biochemical assays.
  • High purity for reproducible experimental results.
  • Available in small research quantities, e.g., 1 mg.
Catalog No. 50-002-36996