Learn More
Medchemexpress LLC Zinterol hydrochloride | 38241-28-0 | 99.7% | C19H27ClN2O4S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14304A1MG
Encompass_Preferred
Zinterol hydrochloride is a potent and selective β2-adrenoceptor agonist. It is used in research to study its effects, such as increasing ICa in a concentration-dependent manner with an EC50 of 2.2 nM. It has been observed to induce ventricular arrhythmias in conscious heart failure rabbits.
  • Potent and selective β2-adrenoceptor agonist.
  • Increases ICa with an EC50 of 2.2 nM.
  • Induces ventricular arrhythmias in conscious heart failure rabbits.
  • For research use only.
Catalog No. 50-003-54427