Learn More
Selleck Chemical LLC ZIP P1083-25MG
SDP
Supplier:  Selleck Chemical LLC P108325MG
Encompass_Preferred
ZIP is a novel cell-permeable protein kinase M (PKM ) inhibitor that blocks PKM -mediated synaptic potentiation
Catalog No. 50-000-42923