Learn More
Apexbio Technology LLC ZIP (SCRAMBLED) 908012-18-0 1mg
SDP
Supplier:  Apexbio Technology LLC A88301
Encompass_Preferred
ZIP SCRAMBLED is a small-molecule inhibitor targeting protein kinase M zeta (PKM ) a constitutive isoform of the protein kinase C (PKC) family It is designed to selectively inhibit PKM thereby modulating the maintenance of late-phase long-term potentiation (L-LTP) a neural mechanism implicated in memory maintenance ZIP SCRAMBLED exerts its biological activity primarily through the inhibition of established L-LTP In experimental settings ZIP SCRAMBLED reverses established L-LTP in hippocampal preparations both in vitro and in vivo Based on these pharmacological properties ZIP SCRAMBLED holds research potential in elucidating PKM -dependent processes underlying memory consolidation and maintenance
Catalog No. 50-000-93870