Learn More
Medchemexpress LLC Ziprasidone hydrochloride | 122883-93-6 | 99.7% | C21H22Cl2N4OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14542A50MG
Encompass_Preferred
Ziprasidone hydrochloride is an orally active combined 5-HT and dopamine receptor antagonist.
  • Acts as a 5-HT and dopamine receptor antagonist
  • Has high affinity for rat D2, 5-HT2A, and 5-HT1A receptors
  • Blocks wild-type hERG current
  • Affects weight gain, physical activity, and thermogenesis in vivo
Catalog No. 50-003-56035