Learn More
Medchemexpress LLC Ziprasidone hydrochloride | 122883-93-6 | MFCD06795476 | 99.7% | C21H22Cl2N4OS | 10MG
SDP
Supplier:  Medchemexpress LLC HY14542A10MG
Encompass_Preferred
Ziprasidone hydrochloride is the hydrochloride salt of ziprasidone (CP-88059), an atypical antipsychotic used as a research reagent. It acts as a combined serotonin (5-HT) and dopamine (D2) receptor antagonist and is supplied as a solid (off-white to light yellow) for pharmacology and biochemical studies. Not for human or veterinary use.
  • High purity suitable for research applications (reported ~99.7%).
  • Well characterized: C21H22Cl2N4OS; CAS 122883-93-6.
  • Supplied in small pack sizes for assay development and screening.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • Relevant for studies of 5-HT2A, 5-HT1A, and D2 receptor pharmacology.
Catalog No. 50-002-61152