Learn More
Medchemexpress LLC Ziprasidone hydrochloride monohydrate | 138982-67-9 | 98.1% | 467.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1740710MM/1ML
Encompass_Preferred
Ziprasidone hydrochloride monohydrate is an orally active combined 5-HT and dopamine receptor antagonist. It shows affinities for Rat D2, 5-HT2A, and 5-HT1A receptors. This product is suitable for various in vitro and in vivo studies.
Catalog No. 50-003-83967