Learn More
Medchemexpress LLC Ziprasidone hydrochloride monohydrate | 138982-67-9 | 98.1% | 467.41 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17407100MG
Encompass_Preferred
Ziprasidone (CP-88059) hydrochloride monohydrate is an orally active combined 5-HT and dopamine receptor antagonist. It exhibits affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM) and 5-HT1A (Ki=3.4 nM) receptors.
  • Orally active combined 5-HT and dopamine receptor antagonist.
  • Affinities for Rat D2, 5-HT2A, and 5-HT1A receptors.
  • Available in solid and solution forms.
Catalog No. 50-003-85779