Learn More
Sino Biological Peptide Substrates: ZIPtide
Supplier:  Sino Biological Z1658
Encompass_Preferred
The ZIPtide substrate peptide sequence (KKLNRTLSFAEPG) is routinely evaluated as a substrate for DAPK3 (ZIPK).
Catalog No. 50-256-7461