Learn More
Strem, An Ascensus Company CAS# 1071-76-7. 250g. Zirconium(IV) n-butoxide (76-80% in n-butanol). MFCD00048765
SDP
Supplier:  Strem, An Ascensus Company 934003250G
Encompass
CAS# 1071-76-7. 250g. Zirconium(IV) n-butoxide (76-80% in n-butanol). MFCD00048765. Molecular Weight: 383.68. Molecular Formula: Zr(OC4H9)4. Color/form: orange liq. Strem# 93-4003. http://www.strem.com/catalog/v/93-4003/
Catalog No. 50-901-15248