Learn More
Cayman Chemical ZIyuglycosIde I 1mg
SDP
Supplier:  Cayman Chemical 373201 MG
Encompass_Preferred
A triterpenoid saponin with diverse biological activities; increases the synthesis of type I collagen of CCD-1064Sk human fibroblast cells; inhibits the proliferation of MDA-MB-231 breast cancer cells (IC50 = 13.96 µM); induces apoptosis in MDA-MB-231 cells and also increases levels of p53 and p21WAF1 in MDA-MB-231 cells
Catalog No. 50-291-7592