Learn More
Medchemexpress LLC ZL0580 | 2377151-10-3 | 99.4% | 532.53 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12642810MG
Encompass_Preferred
ZL0580, an analog of ZL0590, epigenetically suppresses HIV by binding to the BD1 domain of BRD4. It inhibits Tat transactivation and transcription elongation, and also induces a repressive chromatin structure at the HIV promoter.
  • Induces HIV transcriptional suppression.
  • Exhibits low toxicity.
  • Suppresses both PMA-stimulated and basal HIV transcription.
  • Does not cause significant cell death at concentrations below 40 μM.
  • Suppresses HIV in primary CD4+ T cells.
Catalog No. 50-004-36969