Learn More
Medchemexpress LLC ZLC491 | 3053716-46-1 | 98.5% | 866.98 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1681625MG
Encompass_Preferred
ZLC491 is an orally active PROTAC degrader that selectively targets CDK12/CDK13. It induces cereblon- and proteasome-dependent selective degradation of CDK12 and CDK13, inhibiting the transcription and expression of long genes, and mainly acts on a subset of DNA damage response genes. ZLC491 also inhibits the proliferation of various triple-negative breast cancer cells.
  • Selectively targets CDK12/CDK13.
  • Orally active PROTAC degrader.
  • Induces cereblon- and proteasome-dependent degradation.
  • Inhibits transcription and expression of long genes.
  • Acts on a subset of DNA damage response genes.
  • Inhibits proliferation of triple-negative breast cancer cells.
  • Exerts synergistic effect when combined with Cisplatin or Olaparib.
  • Shows extremely low activity against non-cancer cell lines.
  • Oral bioavailability of 46.8%.
Catalog No. 50-003-90846