Learn More
TARGETMOL CHEMICALS INC ZLDI-8 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T134105MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.ZLDI-8 is an inhibitor of Notch activating/cleaving enzyme ADAM-17 and inhibits the cleavage of Notch protein. Purity 98.09%
Catalog No. 50-287-5982