Learn More
Medchemexpress LLC ZLN005-d4 | 2410443-42-2 | 99.5% | 254.36 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17538S1MG
Encompass_Preferred
ZLN005-d4 is a deuterium-labeled version of ZLN005. ZLN005 is a potent activator of peroxisome proliferator-activated receptor-γ coactivator-1α (PGC-1α).
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes, like deuterium, are incorporated into drug molecules and are used as tracers for quantitation during drug development
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-86378