Learn More
Enzo Life Sciences ZM 336372 (5mg). CAS: 208260-29-1
SDP
Supplier:  Enzo Life Sciences BMLEI2980005
Encompass_Preferred
Potent and specific inhibitor of the protein kinase c-Raf (IC50=70nM). Has no significant effect on many other protein kinases tested (even at 50µM) with the exception of p38 (SAPK2a) (IC50=2µM) and SAPK2b (p38β) (IC50=2µM). Induces a paradoxical >100-fold activation of c-Raf in whole cells. Alternative name: N-[5-(3-Dimethylaminobenzamido)-2-methylphenyl]-4-hydroxybenzamide. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (200mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0682