Learn More
Abcam ZM 336372, c-Raf inhibitor, 10MG
Supplier:  Abcam AB12042810MG
Encompass_Preferred
MW 389.4 Da, Purity >98%. Potent, cell-permeable and selective c-Raf inhibitor (IC₅₀ = 70 nM). Displays 10-fold selectivity over b-Raf. Inhibits carcinoid cell growth. Also inhibits growth and apoptosis in pancreatic cancer cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7595