Learn More
Apexbio Technology LLC ZM 39923 HCl 1021868-92-7 50mg
SDP
Supplier:  Apexbio Technology LLC A415150
Encompass_Preferred
ZM 39923 HCl (CAS 1021868-92-7) is a small molecule that selectively inhibits Janus kinases JAK1 and JAK3 displaying pIC50 values of 4 4 and 7 1 respectively As JAK family kinases play a pivotal role in cytokine-mediated signaling via the JAK-STAT pathway ZM 39923 HCl is frequently employed to investigate signal transduction mechanisms linked to hematological malignancies autoimmune conditions and inflammatory disorders The compound also inhibits tissue transglutaminase 2 (TGM2) with an IC50 of 10 nM and exhibits equivalent inhibition of EGFR In cellular studies ZM 39923 HCl modulates T cell proliferation and apoptosis and affects pathways involving VEGF and TGF 2 underscoring its utility in cytokine oncology and cell biology research
Catalog No. 50-000-95347