Learn More
Apexbio Technology LLC ZM 447439, 5mg. Cas: 331771-20-1 MFCD: MFCD08703125. Aurora Kinase inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A41135
Encompass_Preferred
ZM 447439 is a novel, potent and selective inhibitor of Aurora kinase, a family of serine/threonine kinases essential for accurate chromosome segregation during cell division, that inhibits the activity of purified recombinant Aurora A and Aurora B proteins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4884