Learn More
Medchemexpress LLC Zm223 (hydrochloride) | 2438679-27-5 | 98.9% | 538.99 g·mol⁻¹ | C23H18ClF3N4O2S2 | 1 ML

Supplier: Medchemexpress LLC HY101790A10MM 1ML
ZM223 hydrochloride is an orally active, non-covalent inhibitor of the NEDD8-activating enzyme (NAE) with reported anticancer activity. Supplied as solid material and as a ready-to-use 10 mM solution in DMSO (1 mL), the compound is characterized by molecular formula C23H18ClF3N4O2S2, molecular weight 538.99 g·mol⁻¹, and high purity for research use.
- Orally active NAE inhibitor with demonstrated anticancer activity.
- Available as 10 mM solution in DMSO (1 mL) and in multiple solid sizes.
- High purity (98.9%) suitable for biochemical and cellular assays.
- Molecular weight 538.99 g·mol⁻¹; formula C23H18ClF3N4O2S2.
- Supplied as the hydrochloride salt for improved solubility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.