Learn More
TARGETMOL CHEMICALS INC ZN-c3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T96435MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Azenosertib (ZN-c3) is a potent and selective Wee1 inhibitor with balanced potency, ADME, and pharmacokinetic properties.[1] Purity 99.8%
Catalog No. 50-288-0195