Learn More
Aviva Systems Biology Corp ZNF248 Antibody - middle region (ARP39476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF248. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF248. Alternate names: KIAA0137, MGC131882, bA162G10.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57049