Learn More
Aviva Systems Biology Corp ZNF280B Antibody - C-terminal region (ARP39790_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39790_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF280B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF280B. Alternate names: 5'OY11.1, D87009.C22.3, SUHW2, ZNF279, ZNF632ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-014-57180