Learn More
Aviva Systems Biology Corp ZNF286 Antibody - C-terminal region (ARP39424_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39424_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF286. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF286. Alternate names: ZNF286ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57026