Learn More
Aviva Systems Biology Corp ZNF322A Antibody - C-terminal region (ARP39605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF322A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF322A. Alternate names: HCG12, ZNF388, ZNF489, ZNF322AApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53144