Learn More
Aviva Systems Biology Corp ZNF409 Antibody - middle region (ARP32282_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32282_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF409. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF409. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54510