Learn More
Aviva Systems Biology Corp ZNF577 Antibody - C-terminal region (ARP39714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF577. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF577. Alternate names: FLJ32291, MGC4400ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-284-822