Learn More
Aviva Systems Biology Corp ZNF682 Antibody - middle region (ARP39751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF682. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF682. Alternate names: BC39498_3, FLJ90362ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Zebrafish
Catalog No. 50-014-57166