Learn More
Cayman Chemical zngbr offIcnle 6Gngerol 10mg
SDP
Supplier:  Cayman Chemical 1170710 MG
Encompass_Preferred
A natural chemical found in fresh ginger that activates TRPV1 (EC50 = 3.3 µM) as well as the TRP ankyrin receptor TRPA1 (EC50 = 10.4 µM)
Catalog No. 50-295-2355